(S)-2-Amino-N-(4-methyl-2-oxo-2H-chromen-7-yl)propanamide

Supplier: BACHEM AMERICAS INC

4028736.0050 4028736.0250 4028736.1000
I-1410.0050BAEA 211.77 CAD
I-1410.0050BA I-1410.0250BA I-1410.1000BA
(S)-2-Amino-N-(4-methyl-2-oxo-2H-chromen-7-yl)propanamide
(S)-2-Amino-N-(4-methyl-2-oxo-2H-chromen-7-yl)propanamide

Sequence: H-Ala-AMC


Bachem additionally offers deuterated alanine: Fmoc-[D₄]Ala-OH (B-4130).
See also Z-Ala-ONp, C-1040.


Synonym(s): H-Ala-MCA (free base)

Formula: C₁₃H₁₄N₂O₃
Storage Temperature: Freezer
MDL Number: MFCD00077086
CAS Number: 77471-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR