Boc-Phe-Ser-Arg-AMC Acetate
Supplier: BACHEM AMERICAS INC
4012340.0005 4012340.0025
I-1400.0005BAEA 264.44 CAD
I-1400.0005BA I-1400.0025BA
Boc-Phe-Ser-Arg-AMC Acetate
Proteins and Peptides
The fluorogenic substrate Boc-FSR-AMC is preferentially hydrolyzed by coagulation factor XIa. Boc-FSR-AMC is also highly susceptible to the proteases in yeast mitochondrial matrix as well as to tryptase from rat mast cells. In addition, the substrate is cleaved by acrosin from the ascidian Halocynthia roretzi.