H-D-Val-Leu-Lys-AMC Acetate
Supplier: BACHEM AMERICAS INC
4008009.0010 4008009.0050
I-1390.0010BAEA 450.44 CAD
I-1390.0010BA I-1390.0050BA
H-D-Val-Leu-Lys-AMC Acetate
Proteins and Peptides
D-VLK-AMC, sensitive, highly specific fluorescent substrate for plasmin.