H-Val-AMC
Supplier: BACHEM AMERICAS INC
4002284.1000
4031332.0050
4031332.0250
4031332.1000
I-1380.1000BAEA
5036.95
CAD
I-1380.1000BA
I-1385.0050BA
I-1385.0250BA
I-1385.1000BA
H-Val-AMC
H-Val-AMC
Sequence: H-Val-AMC
Bachem additionally offers Fmoc-[¹⁵N]Val-OH, B-2670.
See also Z-Val-ONp, C-2830.
Synonym(s): H-Val-AMC trifluoroacetate salt
Formula:
C₁₅H₁₈N₂O₃ Storage Temperature: Freezer |
MDL Number:
MFCD00080971 CAS Number: 78682-66-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...