H-Tyr-AMC

Supplier: BACHEM AMERICAS INC

4003008.0050 4003008.0250 4003008.1000
I-1375.0050BAEA 180.83 CAD
I-1375.0050BA I-1375.0250BA I-1375.1000BA
H-Tyr-AMC
H-Tyr-AMC

Sequence: H-Tyr-AMC


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.
See also Boc-Tyr-ONp, M-1180.

Formula: C₁₉H₁₈N₂O₄
Storage Temperature: Freezer
MDL Number: MFCD00057306
CAS Number: 94099-57-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR