Tos-Gly-Pro-Arg-AMC · HCl
Supplier: BACHEM AMERICAS INC
4008607.0010
4008607.0050
I-1365.0010BAEA
206.51
CAD
I-1365.0010BA
I-1365.0050BA
Tos-Gly-Pro-Arg-AMC · HCl
Proteins and Peptides
Tos-GPR-AMC, fluorogenic substrate for trypsin and thrombin.