L-Threonine-7-amido-4-methylcoumarin

Supplier: BACHEM AMERICAS INC

Synonyms: H-Thr-AMC

4005542.0050 4005542.0250 4005542.1000
I-1360.0050BAEA 284.04 CAD
I-1360.0050BA I-1360.0250BA I-1360.1000BA
L-Threonine-7-amido-4-methylcoumarin
L-Threonine-7-amido-4-methylcoumarin

Sequence: H-Thr-AMC


Bachem also offers phosphothreonine derivatives.

Formula: C₁₄H₁₆N₂O₄
MW: 276.292 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00152180
CAS Number: 191723-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR