Suc-Leu-Tyr-AMC
Supplier: BACHEM AMERICAS INC
4009168.0050
4009168.0250
I-1355.0050BAEA
313.55
CAD
I-1355.0050BA
I-1355.0250BA
Suc-Leu-Tyr-AMC
Proteins and Peptides
See also I-1115 and I-1395.
Substrate for porcine calpain isozymes I and II and for E.coli protease Ti. Suc-LY-AMC is cleaved by a hyperthermostable subtilisin-type protease bound to the surface layer of the archaeon Staphylothermus marinus.