Suc-Gly-Pro-Leu-Gly-Pro-AMC

Supplier: BACHEM AMERICAS INC

4003958.0025 4003958.0100
I-1350.0025BAEA 903.37 CAD
I-1350.0025BA I-1350.0100BA
Suc-Gly-Pro-Leu-Gly-Pro-AMC
Suc-Gly-Pro-Leu-Gly-Pro-AMC

Sequence: Suc-Gly-Pro-Leu-Gly-Pro-AMC


See also M-2460 and M-2465.


Suc-GPLGP-AMC, a highly sensitive, fluorogenic substrate for thimet oligopeptidase (collagenase-like peptidase, Pz-peptidase, metalloendopeptidase 24.15) as well as for post-proline cleaving enzyme (prolyl endopeptidase).

Formula: C₃₄H₄₄N₆O₁₀
Storage Temperature: Freezer
MDL Number: MFCD00077163
CAS Number: 72698-36-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR