Suc-Ala-Phe-Lys-AMC Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4004022.0050
4031296.0050
I-1325.0050BAEA
1345.09
CAD
I-1325.0050BA
I-1330.0050BA
Suc-Ala-Phe-Lys-AMC Trifluoroacetate
Proteins and Peptides
Suc-AFK-AMC, a highly sensitive fluorogenic substrate for plasmin. Suc-AFK-AMC was also used for assaying gingipain K.