Suc-Ala-Ala-Phe-AMC
Supplier: BACHEM AMERICAS INC
4006306.0050
4006306.0250
I-1315.0050BAEA
774.5
CAD
I-1315.0050BA
I-1315.0250BA
Suc-Ala-Ala-Phe-AMC
Proteins and Peptides
Substrates for measuring the chymotryptic, PGPH, and tryptic acitivity of the 20S or 26S proteasome. See also the thrombin substrate I-1140.
Suc-AAF-AMC, a highly sensitive fluorogenic chymotrypsin substrate. It is also hydrolyzed by neutral metalloendopeptidases, such as thermolysin and enkephalinase (neprilysin, NEP 24.11). Suc-AAF-MCA has also been used for the chymotrypsin-like activity of the 20S proteasome.