Pyr-AMC

Supplier: BACHEM AMERICAS INC

Synonyms: L-Pyroglutamic acid 7-amido-4-methylcoumarin

4004069.0050 4004069.0250 4004069.1000
I-1300.0050BAEA 232.35 CAD
I-1300.0050BA I-1300.0250BA I-1300.1000BA
Pyr-AMC
Pyr-AMC

Sequence: Pyr-AMC


Also called 5-oxoproline or pidolic acid. Bachem uses the acronym Pyr for denoting pyroglutamic acid. Further abbreviations as Glp, Pgl, pGlu (pE), or < Glu (< E) are also in use.


A highly sensitive fluorogenic substrate for pyroglutamyl peptidase I.

Formula: C₁₅H₁₄N₂O₄
Storage Temperature: Freezer
MDL Number: MFCD00080240
CAS Number: 66642-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR