H-Pro-Phe-Arg-AMC Acetate
Supplier: BACHEM AMERICAS INC
4004023.0050 4004023.0250
I-1295.0050BAEA 803.54 CAD
I-1295.0050BA I-1295.0250BA
H-Pro-Phe-Arg-AMC Acetate
Proteins and Peptides
See also I-1160, I-1960, L-1545, M-1130, and M-1140.
A highly sensitive, fluorogenic substrate for plasma as well as pancreatic and urinary kallikreins. PFR-AMC is also cleaved rapidly by soybean trypsin-like enzyme.