(S)-N-(4-Methyl-2-oxo-2H-chromen-7-yl)pyrrolidine-2-carboxamide

Supplier: BACHEM AMERICAS INC

Synonyms: H-Pro-AMC

4001869.0050 4001869.0250 4001869.1000
I-1290.0050BAEA 185.46 CAD
I-1290.0050BA I-1290.0250BA I-1290.1000BA
(S)-N-(4-Methyl-2-oxo-2H-chromen-7-yl)pyrrolidine-2-carboxamide
(S)-N-(4-Methyl-2-oxo-2H-chromen-7-yl)pyrrolidine-2-carboxamide

Sequence: H-Pro-AMC

Formula: C₁₅H₁₆N₂O₃
MW: 272.304 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00213795
CAS Number: 96643-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR