H-Orn-AMC·HCl
Supplier: BACHEM AMERICAS INC
4006789.0250
4006789.1000
I-1280.0250BAEA
1187.51
CAD
I-1280.0250BA
I-1280.1000BA
H-Orn-AMC·HCl
H-Orn-AMC·HCl
Sequence: H-Orn-AMC
Formula:
C₁₅H₁₉N₃O₃·HCl Storage Temperature: Freezer |
MDL Number:
MFCD18633552 CAS Number: 98516-75-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...