Meosuc-Ala-Phe-Lys-AMC·TFA

Supplier: BACHEM AMERICAS INC

Synonyms: MeOSuc-Ala-Phe-Lys-AMC

4006754.0050 4006754.0250
I-1275.0050BAEA 1345.09 CAD
I-1275.0050BA I-1275.0250BA
Meosuc-Ala-Phe-Lys-AMC·TFA
Meosuc-Ala-Phe-Lys-AMC·TFA

Sequence: MeOSuc-Ala-Phe-Lys-AMC


MeOSuc-AFK-AMC, a highly sensitive fluorogenic substrate for plasmin.

Formula: C₃₅H₄₂F₃N₅O₁₀
MW: 749.741 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00152192
CAS Number: 201853-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR