MeOSuc-Ala-Ala-Pro-Val-AMC
Supplier: BACHEM AMERICAS INC
4005227.0050
4005227.0250
I-1270.0050BAEA
774.5
CAD
I-1270.0050BA
I-1270.0250BA
MeOSuc-Ala-Ala-Pro-Val-AMC
Proteins and Peptides
See also C-1040, C-2345, I-1000, and L-1435.
A highly sensitive fluorogenic substrate for the assay of human leukocyte and porcine pancreatic elastases. MeOSuc-AAPV-AMC is also cleaved by myeloblastin.