H-Lys-AMC

Supplier: BACHEM AMERICAS INC

4002577.0250 4002577.1000
I-1255.0250BAEA 1574.38 CAD
I-1255.0250BA I-1255.1000BA
H-Lys-AMC
H-Lys-AMC

Sequence: H-Lys-AMC


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".
See also Z-Lys-ONp · HCl, M-1295.

Formula: C₁₆H₂₁N₃O₃
Storage Temperature: Freezer
MDL Number: MFCD00152123
CAS Number: 92605-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR