L-Leucine(4-methyl-7-coumarinylamide)

Supplier: BACHEM AMERICAS INC

4030806.0250 4030806.1000 4001570.0250 4001570.1000
I-1240.0250BAEA 903.37 CAD
I-1240.0250BA I-1240.1000BA I-1245.0250BA I-1245.1000BA
L-Leucine(4-methyl-7-coumarinylamide)
L-Leucine(4-methyl-7-coumarinylamide)

Sequence: H-Leu-AMC


Bachem additionally offers deuterated and ¹⁵N-labeled leucine: Fmoc-[D₁₀]Leu-OH (B-4120) and Fmoc-[¹⁵N]Leu-OH (B-2655).
Commonly used protease inhibitors as bestatin and the E-64s are derived from leucine. See also H-Leu-NH₂ (E-2650) and Z-Leu-ONp (C-2120).


Synonym(s): H-Leu-MCA hydrochloride salt

Formula: C₁₆H₂₀N₂O₃
MW: 288.347 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00043262
CAS Number: 66447-31-2
EINECS: 266-363-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR