H-Hyp-AMC

Supplier: BACHEM AMERICAS INC

4005319.0250
I-1230.0250BAEA 2284.35 CAD
I-1230.0250BA
H-Hyp-AMC
H-Hyp-AMC

Sequence: H-Hyp-AMC


Hyp denotes L-trans-hydroxyproline.

Formula: C₁₅H₁₆N₂O₄
Storage Temperature: Freezer
MDL Number: MFCD01310881
CAS Number: 77471-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR