Gly pro-7-amido-4-methylcoumarin hydrobromide

Supplier: BACHEM AMERICAS INC

4002520.0025 4002520.0050
I-1225.0025BAEA 678.52 CAD
I-1225.0025BA I-1225.0050BA
Gly pro-7-amido-4-methylcoumarin hydrobromide
Gly pro-7-amido-4-methylcoumarin hydrobromide

Sequence: H-Gly-Pro-AMC · HBr


See also H-Lys-Pro-AMC (I-1745) and H-Lys-Pro-MNA (J-1255) .


Highly sensitive substrate for dipeptidyl aminopeptidase IV (DPP IV, X-prolyldipeptidyl aminopeptidase).

Formula: C₁₇H₂₀BrN₃O₄
Storage Temperature: Freezer
MDL Number: MFCD00152092
CAS Number: 115035-46-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR