Gly-Arg-AMC

Supplier: BACHEM AMERICAS INC

Synonyms: H-Gly-Arg-AMC

4002196.0050 4002196.0250
I-1215.0050BAEA 919.59 CAD
I-1215.0050BA I-1215.0250BA
Gly-Arg-AMC
Gly-Arg-AMC

Sequence: H-Gly-Arg-AMC


Sensitive fluorogenic substrate for dipeptidyl aminopeptidase I (DPP I, cathepsin C).

Formula: C₁₈H₂₄N₆O₄
MW: 388.428 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00155583
CAS Number: 65147-19-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR