2-Amino-N-(4-methyl-2-oxo-2H-chromen-7-yl)acetamide

Supplier: BACHEM AMERICAS INC

4002578.0250 4002578.1000
I-1210.0250BAEA 955.24 CAD
I-1210.0250BA I-1210.1000BA
2-Amino-N-(4-methyl-2-oxo-2H-chromen-7-yl)acetamide
2-Amino-N-(4-methyl-2-oxo-2H-chromen-7-yl)acetamide

Sequence: H-Gly-AMC


Bachem additionally offers deuterated glycine: Fmoc-[D₂]Gly-OH B-4125. For N-methylglycine derivatives see Sarcosine.
See also Boc-Gly-ONp (A-1740) and Z-Gly-ONp (M-1250).

Formula: C₁₂H₁₂N₂O₃
Storage Temperature: Freezer
MDL Number: MFCD00058452
CAS Number: 77471-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR