Glut-Phe-AMC

Supplier: BACHEM AMERICAS INC

Synonyms: Glutaryl-Phe-AMC

4008166.0050 4008166.0250
I-1205.0050BAEA 211.77 CAD
I-1205.0050BA I-1205.0250BA
Glut-Phe-AMC
Glut-Phe-AMC

Sequence: Glutaryl-Phe-AMC


Bachem additionally offers deuterated phenylalanine: Fmoc-[ring-D₅]Phe-OH B-4115.
See also Z-Phe-OH (C-2310) and Z-Phe-ONp (C-2345).


Sensitive fluorogenic substrate for chymotrypsin.

Formula: C₂₄H₂₄N₂O₆
MW: 436.464 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00038083
CAS Number: 58632-47-6
EINECS: 261-369-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR