Glutarylglycylarginine-4-methylcoumaryl-7-amide

Supplier: BACHEM AMERICAS INC

Synonyms: Glutaryl-Gly-Arg-AMC

4005950.0025 4005950.0100
I-1195.0025BAEA 542.13 CAD
I-1195.0025BA I-1195.0100BA
Glutarylglycylarginine-4-methylcoumaryl-7-amide
Glutarylglycylarginine-4-methylcoumaryl-7-amide

Sequence: Glutaryl-Gly-Arg-AMC


Glt-GR-AMC, sensitive fluorogenic substrate for urokinase and tissue-type plasminogen activator.

Formula: C₂₃H₃₀N₆O₇
MW: 502.528 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00058182
CAS Number: 65147-16-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR