H-Gln-AMC

Supplier: BACHEM AMERICAS INC

4003647.0100 4003647.0250
I-1175.0100BAEA 1347.61 CAD
I-1175.0100BA I-1175.0250BA
H-Gln-AMC

Sequence: H-Gln-AMC


For isoglutamine derivatives please see the family "Glutamic Acid".
See also Boc-Gln-ONp (A-1720), H-Gln-OtBu · HCl (E-1920), and Z-Gln-OH (C-1615).


Fluorogenic substrate used in a continuous spectrometric assay for glutaminyl cyclase activity.

Formula: C₁₅H₁₇N₃O₄
Storage Temperature: Freezer
MDL Number: MFCD00237868
CAS Number: 105888-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR