AmbotzHAA7650

Supplier: BACHEM AMERICAS INC

Synonyms: H-Cit-AMC

4006058.0250 4006058.1000
I-1170.0250BAEA 6224.55 CAD
I-1170.0250BA I-1170.1000BA
AmbotzHAA7650
AmbotzHAA7650

Sequence: H-Cit-AMC


Bachem also offers Fmoc-Cit-Wang resin, D-1840.


Synonym(s): H-Orn(carbamoyl)-AMC

Formula: C₁₆H₂₀N₄O₄
MW: 332.36 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00152041
CAS Number: 93753-78-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR