Z-Gly-Pro-Arg-Mca Hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: Z-Gly-Pro-Arg-AMC·HCl

4002047.0025 4002047.0100
I-1150.0025BAEA 715.36 CAD
I-1150.0025BA I-1150.0100BA
Z-Gly-Pro-Arg-Mca Hydrochloride
Z-Gly-Pro-Arg-Mca Hydrochloride

Sequence: Z-Gly-Pro-Arg-AMC · HCl


Z-GPR-AMC is a fluorogenic substrate for cathepsin K, granzyme A, and thrombin.

Formula: C₃₁H₃₈ClN₇O₇
MW: 656.137 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00077020
CAS Number: 201928-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR