Z-Gly-Pro-AMC
Supplier: BACHEM AMERICAS INC
4002518.0050
4002518.0250
I-1145.0050BAEA
461.41
CAD
I-1145.0050BA
I-1145.0250BA
Z-Gly-Pro-AMC
Z-Gly-Pro-AMC
Sequence: Z-Gly-Pro-AMC
See also I-1350 and K-1490.
Z-GP-AMC, fluorogenic substrate for the determination of post-proline cleaving enzyme (prolyl endopeptidase).
Formula:
C₂₅H₂₅N₃O₆ Storage Temperature: Freezer |
MDL Number:
MFCD00038082 CAS Number: 68542-93-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...