Cbz-arg-arg-mca

Supplier: BACHEM AMERICAS INC

Synonyms: Z-Arg-Arg-AMC

4004789.0050 4004789.0250
I-1135.0050BAEA 529.25 CAD
I-1135.0050BA I-1135.0250BA
Cbz-arg-arg-mca
Cbz-arg-arg-mca

Sequence: Z-Arg-Arg-AMC


Z-RR-AMC, an excellent, specific substrate for cathepsin B, an enzyme found in secretory vesicles. Cathepsin B has been identified as a β-secretase, as such it is involved in β-amyloid production. Z-RR-AMC is a convenient fluorogenic substrate for stem bromelain, as it is scarcely affected by fruit bromelain.

Formula: C₃₀H₃₉N₉O₆
MW: 621.699 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00133576
CAS Number: 88937-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR