Z-Arg-AMC

Supplier: BACHEM AMERICAS INC

4001519.0250 4001519.1000
I-1130.0250BAEA 926.8 CAD
I-1130.0250BA I-1130.1000BA
Z-Arg-AMC
Z-Arg-AMC

Sequence: Z-Arg-AMC


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
See also H-Arg(Me)-OH (E-3745) and Fmoc-Arg(Tos)-OH (B-1435).


Sensitive fluorogenic substrate for trypsin, soybean trypsin-like enzyme, and papain.

Formula: C₂₄H₂₇N₅O₅
MW: 465.51 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00039019
CAS Number: 62037-44-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR