Boc-Val-Leu-Lys-AMC Acetate
Supplier: BACHEM AMERICAS INC
4004467.0005 4004467.0025
I-1115.0005BAEA 244.24 CAD
I-1115.0005BA I-1115.0025BA
Boc-Val-Leu-Lys-AMC Acetate
Proteins and Peptides
A sensitive, fluorogenic, and specific substrate for the quantitative measurement of plasmin. Boc-VLK-AMC is also cleaved by acrosin from the ascidian Halocynthia roretzi, by porcine calpain isozymes I and II, and by papain.