Boc-LGR-MCA

Supplier: BACHEM AMERICAS INC

Synonyms: Boc-Leu-Gly-Arg-AMC

4005550.0050 4005550.0250
I-1105.0050BAEA 1086.93 CAD
I-1105.0050BA I-1105.0250BA
Boc-LGR-MCA
Boc-LGR-MCA

Sequence: Boc-Leu-Gly-Arg-AMC


Boc-LGR-AMC, a specific fluorogenic substrate for complement component C3/C5 convertases, coagulation factor Xa, and soybean trypsin-like enzyme. It is also hydrolyzed by macropain, a high molecular weight thiol proteinase from human erythrocytes. Stock solutions of Boc-LGR-AMC are best prepared in DMSO.

Formula: C₂₉H₄₃N₇O₇
Storage Temperature: Freezer
MDL Number: MFCD03788152
CAS Number: 65147-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR