Boc-Ile-Glu-Gly-Arg-AMC Acetate
Supplier: BACHEM AMERICAS INC
4004961.0025
4004961.0100
I-1100.0025BAEA
661.51
CAD
I-1100.0025BA
I-1100.0100BA
Boc-Ile-Glu-Gly-Arg-AMC Acetate
Proteins and Peptides
Boc-IEGR-AMC, a specific, highly fluorogenic substrate for clotting enzyme factor Xa (coagulation factor Xa). Boc-IEGR-AMC is also hydrolyzed by acrosin from the ascidian Halocynthia roretzi. The use of this substrate in the indirect determination of coagulation factor IX has been described.