Boc-Glu-Lys-Lys-AMC

Supplier: BACHEM AMERICAS INC

4007739.0100 4007739.0250
I-1095.0100BAEA 1549.03 CAD
I-1095.0100BA I-1095.0250BA
Boc-Glu-Lys-Lys-AMC
Boc-Glu-Lys-Lys-AMC

Sequence: Boc-Glu-Lys-Lys-AMC


Boc-EKK-AMC, sensitive fluorogenic substrate for urokinase-activated plasmin.

Formula: C₃₂H₄₈N₆O₉
Storage Temperature: Freezer
MDL Number: MFCD00057305
CAS Number: 73554-85-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR