BCMCA
Supplier: BACHEM AMERICAS INC
Synonyms:
H-Cys(Bzl)-AMC
4007281.0050
4007281.0250
I-1090.0050BAEA
439.41
CAD
I-1090.0050BA
I-1090.0250BA
BCMCA
BCMCA
Sequence: H-Cys(Bzl)-AMC
A sensitive fluorogenic substrate for serum cystyl aminopeptidase (oxytocinase).
Formula:
C₂₀H₂₀N₂O₃S MW: 368.451 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00038830 CAS Number: 80173-27-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...