Bz-Val-Gly-Arg-AMC Trifluoroacetate
Supplier: BACHEM AMERICAS INC
Substrates for measuring the chymotryptic, PGPH, and tryptic acitivity of the 20S or 26S proteasome. See also the thrombin substrate I-1140.
Bz-VGR-AMC has been employed as substrate for the tricorn core protease from Thermoplasma acidophilum. Bz-VGR-AMC, I-1085 is used for measuring the trypsin-like activity of the 20S proteasome, in combination with e.g. Suc-LLVY-AMC, I-1395 (for assaying its chymotrypsin-like activity) and Z-LLE-AMC, I-1945 (for determining its PGPH activity).