Bz-Phe-Val-Arg-AMC

Supplier: BACHEM AMERICAS INC

4003131.0005 4003131.0025
I-1080.0005BAEA 244.24 CAD
I-1080.0005BA I-1080.0025BA
Bz-Phe-Val-Arg-AMC
Bz-Phe-Val-Arg-AMC

Sequence: Bz-Phe-Val-Arg-AMC
Synonym(s): Thrombin Substrate III, Fluorogenic


Thrombin substrates are often cleaved as well by trypsin and trypsin-like enzymes, e.g. I-1365, I-1560, J-1120, and L-1150.


Sensitive fluorogenic substrate for the quantitative determination of thrombin. Bz-FVR-AMC is also cleaved by ficain.

Formula: C₃₇H₄₃N₇O₆
MW: 681.794 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00039018
CAS Number: 88899-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR