Nα-Benzoyl-L-arginine-7-amido-4-methylcoumarin hydrochloride

Supplier: BACHEM AMERICAS INC

4002540.0050 4002540.0250
I-1070.0050BAEA 217 CAD
I-1070.0050BA I-1070.0250BA
Nα-Benzoyl-L-arginine-7-amido-4-methylcoumarin hydrochloride
Nalpha-Benzoyl-L-arginine-7-amido-4-methylcoumarin hydrochloride

Sequence: Bz-Arg-AMC · HCl


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
See also H-Arg(Me)-OH (E-3745) and Fmoc-Arg(Tos)-OH (B-1435).


Sensitive fluorogenic substrate for trypsin, soybean trypsin-like enzyme, and papain.

Formula: C₂₃H₂₆ClN₅O₄
Storage Temperature: Freezer
MDL Number: MFCD00039020
CAS Number: 83701-04-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR