H-Asp(AMC)-OH

Supplier: BACHEM AMERICAS INC

4005538.0250 4005538.1000
I-1060.0250BAEA 4339.91 CAD
I-1060.0250BA I-1060.1000BA
H-Asp(AMC)-OH
H-Asp(AMC)-OH

Sequence: H-Asp(AMC)-OH


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.


Substrate for a sensitive and specific fluorometric assay of glycosylasparaginase activity. This assay is useful for the rapid diagnosis of aspartylglycosaminuria, the most common lysosomal storage disorder of glycoprotein degradation. Also used for assaying β-aspartyl dipeptidase.

Formula: C₁₄H₁₄N₂O₅
MW: 290.28 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00236804
CAS Number: 133628-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR