H-Arg-AMC

Supplier: BACHEM AMERICAS INC

4002148.0250 4002148.1000
I-1050.0250BAEA 1383.8 CAD
I-1050.0250BA I-1050.1000BA
H-Arg-AMC
H-Arg-AMC

Sequence: H-Arg-AMC


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
See also H-Arg(Me)-OH (E-3745) and Fmoc-Arg(Tos)-OH (B-1435).


Sensitive fluorogenic substrate for a convenient assay of cathepsin H. It is also cleaved by aminopeptidase B (arginine aminopeptidase).

Formula: C₁₆H₂₁N₅O₃
MW: 331.38 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00058184
CAS Number: 70274-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR