H-D-Ala-Leu-Lys-AMC

Supplier: BACHEM AMERICAS INC

4004459.0005 4004459.0025
I-1040.0005BAEA 258.14 CAD
I-1040.0005BA I-1040.0025BA
H-D-Ala-Leu-Lys-AMC
H-D-Ala-Leu-Lys-AMC

Sequence: H-D-Ala-Leu-Lys-AMC


ALK-AMC, sensitive, highly specific fluorogenic substrate for plasmin.

Formula: C₂₅H₃₇N₅O₅
Storage Temperature: Freezer
MDL Number: MFCD18782512
CAS Number: 104881-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR