3-Phenyl-L-serine
Supplier: BACHEM AMERICAS INC
Synonyms:
L-threo-Phenylserine
4064845.0001
4064845.0005
F-4225.0001BAEA
461.41
CAD
F-4225.0001BA
F-4225.0005BA
3-Phenyl-L-serine
3-Phenyl-L-serine
Sequence: L-threo-Phenylserine
Synonym(s): (2S,3R)-2-Amino-3-hydroxy-3-phenylpropionic acid
A building block of peptide antibiotics and a valuable chiral synthon. Crich and Banerjee used it for obtaining β-mercaptophenylalanine. Introduction of this Phe derivative allowed native chemical ligation at Phe followed by desulfuration.
Formula:
C₉H₁₁NO₃ MW: 181.191 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00671668 CAS Number: 6254-48-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...