(S)-methyl-2-aminobutanoate

Supplier: BACHEM AMERICAS INC

Synonyms: H-Abu-OMe·HCl

4049719.0005 4049719.0025
F-4190.0005BAEA 645.43 CAD
F-4190.0005BA F-4190.0025BA
(S)-methyl-2-aminobutanoate
(S)-methyl-2-aminobutanoate

Sequence: H-Abu-OMe · HCl

Formula: C₅H₁₁NO₂
MW: 117.148 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09037399
CAS Number: 15399-22-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR