(R)-α-Methylleucine

Supplier: BACHEM AMERICAS INC

Synonyms: H-D-α-Me-Leu-OH, D-Leucine, 2-methyl-, (R)-2-Amino-2,4-dimethylpentanoic acid, H-α-Me-D-Leu-OH

4047055.0001 4047055.0005
F-4155.0001BAEA 774.5 CAD
F-4155.0001BA F-4155.0005BA
(R)-α-Methylleucine
(R)-alpha-Methylleucine

Sequence: H-α-Me-D-Leu-OH


Bachem additionally offers deuterated and ¹⁵N-labeled leucine: Fmoc-[D₁₀]Leu-OH (B-4120) and Fmoc-[¹⁵N]Leu-OH (B-2655).

Formula: C₇H₁₅NO₂
MW: 145.202 g/mol
Storage Temperature: Freezer
MDL Number: MFCD08752476
CAS Number: 29589-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR