L(+)-Citrulline

Supplier: BACHEM AMERICAS INC

Synonyms: H-Cit-OH

4031894.0100 4031894.0250
F-4145.0100BAEA 321.59 CAD
F-4145.0100BA F-4145.0250BA
L(+)-Citrulline
L(+)-Citrulline

Sequence: H-Cit-OH


Bachem also offers Fmoc-Cit-Wang resin, D-1840.


Synonym(s): H-Orn(carbamoyl)-OH

Formula: C₆H₁₃N₃O₃
Storage Temperature: Ambient
MDL Number: MFCD00064397
CAS Number: 372-75-8
EINECS: 206-759-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR