N-ME-D-PHG-OH

Supplier: BACHEM AMERICAS INC

4038768.0001 4038768.0005
F-4140.0001BAEA 284.04 CAD
F-4140.0001BA F-4140.0005BA
N-ME-D-PHG-OH
N-ME-D-PHG-OH

Sequence: N-Me-D-Phg-OH

Formula: C₉H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD08458661
CAS Number: 30925-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR