H-D-Abu-OtBu·HCl

Supplier: BACHEM AMERICAS INC

Synonyms: H-D-Abu-OtBu·HCl

4042844.0001 4042844.0005
F-4110.0001BAEA 516.34 CAD
F-4110.0001BA F-4110.0005BA
H-D-Abu-OtBu·HCl
H-D-Abu-OtBu·HCl

Sequence: H-D-Abu-OtBu · HCl

Formula: C₈H₁₈ClNO₂
MW: 195.687 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08275809
CAS Number: 313994-32-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR