(R)-2-Hydroxy-4-methylpentanoic acid
Supplier: BACHEM AMERICAS INC
Synonyms:
D-α-Hydroxyisocaproic acid, (R)-Leucic acid
4026246.0001
4026246.0005
F-4050.0001BAEA
258.14
CAD
F-4050.0001BA
F-4050.0005BA
(R)-2-Hydroxy-4-methylpentanoic acid
(R)-2-Hydroxy-4-methylpentanoic acid
Sequence: D-α-Hydroxyisocaproic acid
Useful chiral synthons, building blocks for depsipeptides.
Synonym(s): D-Leucic acid
Formula:
C₆H₁₂O₃ MW: 132.159 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00211263 CAS Number: 20312-37-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...