Benzyl-4-aminobutanoate-4-methylbenzenesulfonate
Supplier: BACHEM AMERICAS INC
Synonyms:
H-γ-Abu-OBzl·p-tosylate, H-γ-Abu-Obzl.TosOH, H-γ-Abu-OBzl·TosOH
4011374.0025
4011374.0100
F-4045.0025BAEA
258.14
CAD
F-4045.0025BA
F-4045.0100BA
Benzyl-4-aminobutanoate-4-methylbenzenesulfonate
Benzyl-4-aminobutanoate-4-methylbenzenesulfonate
Sequence: H-γ-Abu-OBzl · p-tosylate
Flexible spacer molecules of various length.
Synonym(s): H-GABA-OBzl · p-tosylate
Formula:
C₁₈H₂₃NO₅S MW: 365.444 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03424226 CAS Number: 26727-22-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...